Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,20-22mm-48P 6x8 Spacing
SDP
Catalog No. 501945315
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 1 1/16" (26.9mm) Tube size: 25mm Spacing: 6x8 holds 48 tubes Dimensions: 8 1/4" X 6 3/16"X 3 1/2" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5315 Supplier Bellco Glass, Inc. Supplier No. 202720048
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 1 1/16" (26.9mm) Tube size: 25mm Spacing: 6x8 holds 48 tubes Dimensions: 8 1/4" X 6 3/16"X 3 1/2" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each