Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,25mm-40P 4x10 Spacing
SDP
Catalog No. 501945318
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 1 1/16" (26.9mm) Tube size: 25mm Spacing:4x10 holds 40 tubes Dimensions: 11 1/2" x 4 3/4" x 4" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .162"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5318 Supplier Bellco Glass, Inc. Supplier No. 202725040
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 1 1/16" (26.9mm) Tube size: 25mm Spacing:4x10 holds 40 tubes Dimensions: 11 1/2" x 4 3/4" x 4" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .162"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each