Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,38mm-24P 4x6 Spacing
SDP
Catalog No. 501945322
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 1 9/16" (39.6mm) Tube size: 38mm Spacing: 4x6 holds 24 tubes Dimensions: 10" x 6 3/4" x 5" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .162"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5322 Supplier Bellco Glass, Inc. Supplier No. 202738024
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 1 9/16" (39.6mm) Tube size: 38mm Spacing: 4x6 holds 24 tubes Dimensions: 10" x 6 3/4" x 5" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .162"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each