Learn More
Bellco Glass, Inc. Stainless Steel Ultra-Vented Inc Shelf For Bellco Roll-in Inc
SDP
Catalog No. 501946052
Encompass_Preferred
Approx 25 x 34" (63.5 x 86.5cm).Stainless steel mesh with reinforcing bars.. MADE IN THE USAFits on adjustable clips inside incubator.Load: 240 lbs (max).
Catalog No. 50-194-6052 Supplier Bellco Glass, Inc. Supplier No. 772840050
Approx 25 x 34" (63.5 x 86.5cm).Stainless steel mesh with reinforcing bars.. MADE IN THE USAFits on adjustable clips inside incubator.Load: 240 lbs (max).