Learn More
Bellco Glass, Inc. Stainless Steel Vertical Disk Impeller,10L For 10L Bioreactor Systems
SDP
Catalog No. 501944730
Encompass_Preferred
SST Vertical Disk Impeller for 10l bioreactor systems - English threads MADE IN THE USAMaterial: 316l SST with electropolish finish3 1/4" od
Catalog No. 50-194-4730 Supplier Bellco Glass, Inc. Supplier No. 196402100
SST Vertical Disk Impeller for 10l bioreactor systems - English threads MADE IN THE USAMaterial: 316l SST with electropolish finish3 1/4" od