Learn More
Eurofins Discoverx Ready-to-Assay SST5 Somatostatin Receptor Frozen Cells, 2 Vials of Cells. (HTS139RTA)
Catalog No. 502005962
Encompass
The Ready-to-Assay SST5 Somatostatin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. somatostatin) based activation of SSTR5 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses SSTR5 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of SSTR5, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5962 Supplier Eurofins Discoverx Supplier No. HTS139RTA
The Ready-to-Assay SST5 Somatostatin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. somatostatin) based activation of SSTR5 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses SSTR5 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of SSTR5, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.