Learn More
Eurofins Discoverx PathHunter SSTR2 Bioassay Kit, 10-Plate. (93-0181Y2-00068)
Catalog No. 501999884
Encompass
The PathHunter SSTR2 Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. somatostatin 28) based activation of SSTR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SSTR2 and EA-tagged β-Arrestin-2. Activation of SSTR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-199-9884 Supplier Eurofins Discoverx Supplier No. 930181Y200068
Add to Cart
Add to Cart
Request a Quote
The PathHunter SSTR2 Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. somatostatin 28) based activation of SSTR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SSTR2 and EA-tagged β-Arrestin-2. Activation of SSTR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.