Learn More
Aviva Systems Biology Corp SSTR4 Antibody - middle region (ARP33359_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33359_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSTR4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SSTR4. Alternate names: SS4R, SS4-R, SS-4-RApplicationWB. Species reactivity: Mouse, Rabbit
Catalog No. 50-014-54979