Learn More
Medchemexpress LLC HY-102037 10mg Medchemexpress, SSTR5 antagonist 1 CAS:1628741-91-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10203710MG
Encompass_Preferred
Medchemexpress, HY-102037 10mg SSTR5 antagonist 1 CAS:1628741-91-2 SSTR5 antagonist 1 is a potent, selective, and orally available somatostatin receptor subtype 5 (SSTR5) antagonist with IC50s of 9.6 and 57 nM for hSSTR5 and mSSTR5, respectively. (Compound 25a) Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7963