Learn More
Aviva Systems Biology Corp SSU72 Antibody - C-terminal region (ARP64592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSU72. It was validated on Western Blot. . Alternate names: FLJ13048, HSPC182, PNAS-120ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-305