Learn More
Perkin Elmer US LLC S-Swafer Complete Kit for Clarus with PPC
Catalog No. 502117119
Encompass_Preferred
Swafer micro-channel wafer technology is an innovative and user-friendly approach for flowswitching and splitting applications - it delivers unparalleled hardware and application flexibility, expanding the capabilities of capillary gas chromatography (GC).
Catalog No. 50-211-7119 Supplier Perkin Elmer US LLC Supplier No. N6520272
Add to Cart
Add to Cart
Request a Quote
Swafer micro-channel wafer technology is an innovative and user-friendly approach for flowswitching and splitting applications - it delivers unparalleled hardware and application flexibility, expanding the capabilities of capillary gas chromatography (GC).