Learn More
Perkin Elmer US LLC S-Swafer Splitter
Catalog No. 502112912
Encompass_Preferred
Swafer micro-channel wafer technology is an innovative and user-friendly approach for flowswitching and splitting applications - it delivers unparalleled hardware and application flexibility, expanding the capabilities of capillary gas chromatography (GC).
Catalog No. 50-211-2912 Supplier Perkin Elmer US LLC Supplier No. N9306262
Swafer micro-channel wafer technology is an innovative and user-friendly approach for flowswitching and splitting applications - it delivers unparalleled hardware and application flexibility, expanding the capabilities of capillary gas chromatography (GC).