Learn More
Aviva Systems Biology Corp SSX2 Antibody - C-terminal region (ARP37719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SSX2. Alternate names: HD21, HOM-MEL-40, MGC119055, MGC15364, MGC3884, SSX, SSX2B, CT5.2aApplicationWB. Species reactivity: Human
Catalog No. 50-014-56406