Learn More
Aviva Systems Biology Corp SSX2IP Antibody - middle region (ARP53690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSX2IP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SSX2IP. Alternate names: ADIP, FLJ10848, KIAA0923, MGC75026ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61092