Learn More
Aviva Systems Biology Corp SSX3 Antibody - N-terminal region (ARP39469_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39469_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSX3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SSX3. Alternate names: CT5.3, MGC119054, MGC14495ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-809