Learn More
Aviva Systems Biology Corp SSX4 Antibody - middle region (ARP35783_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35783_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SSX4. Alternate names: MGC119056, MGC12411, CT5.4, SSX4BApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-55847