Learn More
Aviva Systems Biology Corp SSX5 Antibody - N-terminal region (ARP39470_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39470_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSX5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SSX5. Alternate names: MGC9494ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57047