Learn More
Aviva Systems Biology Corp SSX6P Antibody - middle region (ARP36225_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36225_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSX6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SSX6. Alternate names: dJ54B20.1, SSXP2, psiSSX2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53040