Learn More
TARGETMOL CHEMICALS INC ST-1006 5MG
SDP
Catalog No. 502580248
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ST-1006 is a potent histamine H4 receptor agonist (pKi 7.94).ST-1006 has anti-inflammatory effects.ST-1006 is a potent inducer of basophil migration and induces basophil migration.ST-1006 is a potent inducer of basophil migration. ST-1006 has anti-inflammatory and anti-pruritic effects. purity: 99%
Catalog No. 50-258-0248 Supplier TARGETMOL CHEMICALS INC Supplier No. T347095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ST-1006 is a potent histamine H4 receptor agonist (pKi 7.94).ST-1006 has anti-inflammatory effects.ST-1006 is a potent inducer of basophil migration and induces basophil migration.ST-1006 is a potent inducer of basophil migration. ST-1006 has anti-inflammatory and anti-pruritic effects. purity: 99%