Learn More
Medchemexpress LLC ST-148 maleate | 390803-40-4 | 99.4% | 612.74 | 1 MG
SDP
Catalog No. 5000348696
Encompass_Preferred
ST-148 maleate is a potent and orally active DENV inhibitor. It demonstrates antiviral efficacy and low cell toxicity. ST-148 maleate works by altering the interaction between lipid droplets and the C protein, which in turn inhibits viral replication.
  • Potent and orally active DENV inhibitor
  • Demonstrates antiviral efficacy
  • Low cell toxicity
  • Inhibits viral replication
Catalog No. 50-003-48696 Supplier Medchemexpress LLC Supplier No. HY1367821MG
ST-148 maleate is a potent and orally active DENV inhibitor. It demonstrates antiviral efficacy and low cell toxicity. ST-148 maleate works by altering the interaction between lipid droplets and the C protein, which in turn inhibits viral replication.
  • Potent and orally active DENV inhibitor
  • Demonstrates antiviral efficacy
  • Low cell toxicity
  • Inhibits viral replication