Learn More
Medchemexpress LLC St-148 maleate | 390803-40-4 | 99.4% | 612.74 g/mol | C31H40N4O7S | 5 MG
SDP
Catalog No. 5000222408
Encompass_Preferred
ST-148 maleate is a research-grade small-molecule inhibitor of dengue virus (DENV). It is reported to be orally active, exhibits antiviral efficacy with low cellular toxicity, and inhibits viral replication by altering interactions between lipid droplets and the viral C protein. Supplied as a 5 mg research sample for biochemical and virology applications.
  • Potent orally active DENV inhibitor.
  • Alters lipid droplet-C protein interaction to block replication.
  • High purity suitable for experimental use (reported 99.4%).
  • Small 5 mg sample for assay development and screening.
  • Molecular formula C31H40N4O7S; molecular weight 612.74 g/mol.
Catalog No. 50-002-22408 Supplier Medchemexpress LLC Supplier No. HY1367825MG
ST-148 maleate is a research-grade small-molecule inhibitor of dengue virus (DENV). It is reported to be orally active, exhibits antiviral efficacy with low cellular toxicity, and inhibits viral replication by altering interactions between lipid droplets and the viral C protein. Supplied as a 5 mg research sample for biochemical and virology applications.
  • Potent orally active DENV inhibitor.
  • Alters lipid droplet-C protein interaction to block replication.
  • High purity suitable for experimental use (reported 99.4%).
  • Small 5 mg sample for assay development and screening.
  • Molecular formula C31H40N4O7S; molecular weight 612.74 g/mol.