Learn More
Medchemexpress LLC HY-101441A 5mg Medchemexpress, ST-193 (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501932131
Encompass_Preferred
Medchemexpress, HY-101441A 5mg ST-193 (hydrochloride) CAS: ST-193 hydrochloride is a potent broad-spectrum arenavirus inhibitor; inhibits Guanarito, Junin, Lassa and Machupo virus with IC 50 values of 0.44, 0.62, 1.4 and 3.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2131 Supplier Medchemexpress LLC Supplier No. HY101441A5MG
Medchemexpress, HY-101441A 5mg ST-193 (hydrochloride) CAS: ST-193 hydrochloride is a potent broad-spectrum arenavirus inhibitor; inhibits Guanarito, Junin, Lassa and Machupo virus with IC 50 values of 0.44, 0.62, 1.4 and 3.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.