Learn More
Medchemexpress LLC St-193 hydrochloride | 2320274-72-2 | 98.8% | 407.94 g/mol | C24H26ClN3O | 1 ML
SDP
Catalog No. 5000245768
Encompass_Preferred
ST-193 hydrochloride is a potent, broad-spectrum arenavirus inhibitor for in vitro antiviral research. It exhibits low-nanomolar activity against Guanarito (IC50 0.44 nM), Junin (0.62 nM), Lassa (1.4 nM), and Machupo (3.1 nM). The compound is supplied as a defined solution with documented purity and physicochemical data for reproducible experimental use.
  • Low-nanomolar inhibition of multiple arenaviruses.
  • Suitable for in vitro antiviral assay development and profiling.
  • High documented purity for consistent results.
  • Available as a ready-to-use solution to simplify dosing.
  • Soluble in DMSO; limited water solubility, consistent with guidelines for handling.
Catalog No. 50-002-45768 Supplier Medchemexpress LLC Supplier No. HY101441A10MM 1ML
ST-193 hydrochloride is a potent, broad-spectrum arenavirus inhibitor for in vitro antiviral research. It exhibits low-nanomolar activity against Guanarito (IC50 0.44 nM), Junin (0.62 nM), Lassa (1.4 nM), and Machupo (3.1 nM). The compound is supplied as a defined solution with documented purity and physicochemical data for reproducible experimental use.
  • Low-nanomolar inhibition of multiple arenaviruses.
  • Suitable for in vitro antiviral assay development and profiling.
  • High documented purity for consistent results.
  • Available as a ready-to-use solution to simplify dosing.
  • Soluble in DMSO; limited water solubility, consistent with guidelines for handling.