Learn More
Medchemexpress LLC HY-50937 1mg Medchemexpress, ST 2825 CAS:894787-30-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY509371MG
Encompass_Preferred
Medchemexpress, HY-50937 1mg ST 2825 CAS:894787-30-5 ST 2825 is a specific MyD88 dimerization inhibitor. ST2825 interferes with recruitment of IRAK1 and IRAK4 by MyD88, causing inhibition of IL-1β-mediated activation of NF-κB transcriptional activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9356
 Restricted Item