Learn More
Medchemexpress LLC ST-836 hydrochloride | 1415564-68-9 | C23H35ClN4OS | 1 ML
SDP
Catalog No. 5000372600
Encompass_Preferred
ST-836 hydrochloride is a potent dopamine receptor ligand with potential for Parkinson's disease research. It exhibits Ki values of 4.5 nM for D3 and 132 nM for D2 receptors. This compound is provided as a 10 mM solution in dimethyl sulfoxide (DMSO).
  • Potent dopamine receptor ligand
  • Potential for Parkinson's disease research
  • Enhanced water solubility and stability in salt form
Catalog No. 50-003-72600 Supplier Medchemexpress LLC Supplier No. HY15238A10MM/1ML
ST-836 hydrochloride is a potent dopamine receptor ligand with potential for Parkinson's disease research. It exhibits Ki values of 4.5 nM for D3 and 132 nM for D2 receptors. This compound is provided as a 10 mM solution in dimethyl sulfoxide (DMSO).
  • Potent dopamine receptor ligand
  • Potential for Parkinson's disease research
  • Enhanced water solubility and stability in salt form