Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ST-836 (hydrochloride) | 1415564-68-9 | C23H35ClN4OS | 25 MG
SDP

Catalog No. 5000374586
Encompass_Preferred

ST-836 hydrochloride is a potent dopamine receptor ligand with Ki values of 4.5 nM and 132 nM for D3 and D2 receptors, respectively. It has potential for Parkinson's disease. The salt form typically offers enhanced water solubility and stability compared to the free form, while maintaining comparable biological activity at equivalent molar concentrations.

  • Potent dopamine receptor ligand
  • Targets D3 receptor (Ki: 4.5 nM)
  • Targets D2 receptor (Ki: 132 nM)
  • Potential for Parkinson's disease treatment
  • Enhanced water solubility and stability (salt form)
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture

Catalog No. 50-003-74586 Supplier Medchemexpress LLC Supplier No. 25MGHY15238A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ST-836 hydrochloride is a potent dopamine receptor ligand with Ki values of 4.5 nM and 132 nM for D3 and D2 receptors, respectively. It has potential for Parkinson's disease. The salt form typically offers enhanced water solubility and stability compared to the free form, while maintaining comparable biological activity at equivalent molar concentrations.

  • Potent dopamine receptor ligand
  • Targets D3 receptor (Ki: 4.5 nM)
  • Targets D2 receptor (Ki: 132 nM)
  • Potential for Parkinson's disease treatment
  • Enhanced water solubility and stability (salt form)
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.