Learn More
TARGETMOL CHEMICALS INC ST 91 5MG
SDP
Catalog No. 502580874
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ST-91 is an agonist of (alpha)2-adrenoceptor with a mixed (alpha)-adrenergic receptor type/subtype selection profile. purity: 99%
Catalog No. 50-258-0874 Supplier TARGETMOL CHEMICALS INC Supplier No. T233975MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ST-91 is an agonist of (alpha)2-adrenoceptor with a mixed (alpha)-adrenergic receptor type/subtype selection profile. purity: 99%