Learn More
Accuflow Systems Saint-Gobain PharmaPure Peristaltic Tubing, 2.4 mm I.D., 5.5 mm O.D.
Catalog No. NC2884903
Encompass_Preferred
PharmaPure low-spallation peristaltic pump tubing engineered for bioprocess and aseptic fluid transfer. It provides improved durability versus silicone, low permeability, and validated biocompatibility for cell culture, vaccine, and diagnostic workflows.
  • 3/32 in (2.4 mm) inner diameter, 7/32 in (5.5 mm) outer diameter.
  • Wall thickness 1/16 in (1.6 mm).
  • Autoclavable; ethylene oxide and gamma irradiation compatible (25 kGy).
  • Maximum service temperature 275 °F (135 °C); low temperature embrittlement -89 °F (-67 °C).
  • Low spallation, low permeability, reduced absorption.
  • Suitable for cell harvest, vaccine manufacturing, bioreactor lines, aseptic filling, diagnostics, and filtration processes.
Catalog No. NC2884903 Supplier Accuflow Systems Supplier No. AL242005
PharmaPure low-spallation peristaltic pump tubing engineered for bioprocess and aseptic fluid transfer. It provides improved durability versus silicone, low permeability, and validated biocompatibility for cell culture, vaccine, and diagnostic workflows.
  • 3/32 in (2.4 mm) inner diameter, 7/32 in (5.5 mm) outer diameter.
  • Wall thickness 1/16 in (1.6 mm).
  • Autoclavable; ethylene oxide and gamma irradiation compatible (25 kGy).
  • Maximum service temperature 275 °F (135 °C); low temperature embrittlement -89 °F (-67 °C).
  • Low spallation, low permeability, reduced absorption.
  • Suitable for cell harvest, vaccine manufacturing, bioreactor lines, aseptic filling, diagnostics, and filtration processes.