Learn More
Accuflow Systems Saint-Gobain Tygon Laboratory Tubing, Non-DEHP, 1/16 in ID
Catalog No. 502365765
Encompass_Preferred
Tygon E-3603 laboratory tubing is a clear, flexible, non-DEHP PVC tubing formulated for laboratory fluid transfer, peristaltic pump use, and vacuum applications. It features extra-heavy walls for vacuum resistance, a smooth polished inner wall to minimize buildup, and high flex-fatigue and abrasion resistance for extended service life.
  • 1/16 in inner diameter, 1/8 in outer diameter, 1/32 in wall thickness.
  • 500 ft (152.4 m) length per roll.
  • Clear, non-DEHP PVC, phthalate-free.
  • Withstands full vacuum at 73°F (29.9 inHg / 760 mmHg).
  • Maximum recommended operating temperature 165°F (74°C).
  • Resistant to flex-fatigue and abrasion for pump and instrument lines.
Catalog No. 50-236-5765 Supplier Accuflow Systems Supplier No. ACF04002
Tygon E-3603 laboratory tubing is a clear, flexible, non-DEHP PVC tubing formulated for laboratory fluid transfer, peristaltic pump use, and vacuum applications. It features extra-heavy walls for vacuum resistance, a smooth polished inner wall to minimize buildup, and high flex-fatigue and abrasion resistance for extended service life.
  • 1/16 in inner diameter, 1/8 in outer diameter, 1/32 in wall thickness.
  • 500 ft (152.4 m) length per roll.
  • Clear, non-DEHP PVC, phthalate-free.
  • Withstands full vacuum at 73°F (29.9 inHg / 760 mmHg).
  • Maximum recommended operating temperature 165°F (74°C).
  • Resistant to flex-fatigue and abrasion for pump and instrument lines.