Learn More
Bellco Glass, Inc. Stainless Steel Harvesting Pipe Assembly , 7L Straight - For 10mm Port
SDP
Catalog No. 501946444
Encompass_Preferred
Straight Harvesting Pipe Assembly 7L. Used for withdraw of contentsfrom 3L bioreactor vessel. Fits 10mm port on 3L metric headplate. MADE IN THE USA
Catalog No. 50-194-6444 Supplier Bellco Glass, Inc. Supplier No. 790900035
Straight Harvesting Pipe Assembly 7L. Used for withdraw of contentsfrom 3L bioreactor vessel. Fits 10mm port on 3L metric headplate. MADE IN THE USA