Learn More
Medchemexpress LLC St-Ht31 (ammonium) | 96.6% | 2798.32 (free base) | 1 MG
SDP
Catalog No. 5000424029
Encompass_Preferred
St-Ht31 ammonium is a membrane-permeable peptide that inhibits protein kinase A (PKA) anchoring. It facilitates cholesterol/phospholipid efflux, which can completely reverse foam cell formation and restore the metabolic health of macrophages.
  • Membrane-permeable peptide inhibitor of PKA anchoring.
  • Induces robust cholesterol/phospholipid efflux.
  • Completely reverses foam cell formation.
  • Restores the metabolic health of macrophages.
Catalog No. 50-004-24029 Supplier Medchemexpress LLC Supplier No. HYP2624A1MG
St-Ht31 ammonium is a membrane-permeable peptide that inhibits protein kinase A (PKA) anchoring. It facilitates cholesterol/phospholipid efflux, which can completely reverse foam cell formation and restore the metabolic health of macrophages.
  • Membrane-permeable peptide inhibitor of PKA anchoring.
  • Induces robust cholesterol/phospholipid efflux.
  • Completely reverses foam cell formation.
  • Restores the metabolic health of macrophages.