Learn More
Cayman Chemical st-Ht31 (ammonium salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502909276
Encompass_Preferred
A cell-permeable stearated peptide; decreases PKA membrane anchoring but not activity in a reporter assay using BHK-21 fibroblasts at 50 µM; reduces capacitance, hyperactivation, as well as prevents in vitro fertilization, in mouse sperm at 1 and 3 µM; increases cholesterol export from ABCA1-expressing BHK-21 cells and RAW 264.7 macrophages at 50 µM
Catalog No. 50-290-9276 Supplier Cayman Chemical Supplier No. 367455 MG
A cell-permeable stearated peptide; decreases PKA membrane anchoring but not activity in a reporter assay using BHK-21 fibroblasts at 50 µM; reduces capacitance, hyperactivation, as well as prevents in vitro fertilization, in mouse sperm at 1 and 3 µM; increases cholesterol export from ABCA1-expressing BHK-21 cells and RAW 264.7 macrophages at 50 µM