Learn More
BioVendor, LLC QuickZyme Sensitive Tissue Hydroxyproline Kit 96-Assays (for collagen analysis)
SDP
Catalog No. 502069129
Encompass
QuickZyme Sensitive Tissue Hydroxyproline Kit, 96-Assays. Measure hydroxyproline in small tissue samples with low collagen levels! The ONLY hydroxyproline-based assay which shows no matrix effect and no need for further dilution of the hydrolysate, enabling measurement in small samples with low hydroxyproline levels. No drying of hydrolysate is needed, resulting in a fast (<2 hrs), sensitive and easy (96-well plate format) assay. Dynamic range: 6-300 uM. Sensitivity: 0.3 ug/ml. Species independent. Measures total hydroxyproline. If this hydroxyproline is obtained upon hydrolysis of collagen, it represents all collagen types present without discriminating between types of collagen, procollagen, mature collagen and collagen degradation products. Hydroxyproline in tissue hydrolysates can be used as a direct measure of the amount of collagen present. Collagen hydrolysis tubes & protocol not included (if these items are needed, please order QuickZyme Sensitive Tissue Collagen Assay).
Catalog No. 50-206-9129 Supplier BioVendor, LLC Supplier No. QZBTISHYP1
QuickZyme Sensitive Tissue Hydroxyproline Kit, 96-Assays. Measure hydroxyproline in small tissue samples with low collagen levels! The ONLY hydroxyproline-based assay which shows no matrix effect and no need for further dilution of the hydrolysate, enabling measurement in small samples with low hydroxyproline levels. No drying of hydrolysate is needed, resulting in a fast (<2 hrs), sensitive and easy (96-well plate format) assay. Dynamic range: 6-300 uM. Sensitivity: 0.3 ug/ml. Species independent. Measures total hydroxyproline. If this hydroxyproline is obtained upon hydrolysis of collagen, it represents all collagen types present without discriminating between types of collagen, procollagen, mature collagen and collagen degradation products. Hydroxyproline in tissue hydrolysates can be used as a direct measure of the amount of collagen present. Collagen hydrolysis tubes & protocol not included (if these items are needed, please order QuickZyme Sensitive Tissue Collagen Assay).