Learn More
Leica Biosystems ST INFINITY H E STAINING SYTEM
Catalog No. NC1231542
Encompass_Preferred
All stains and reagents are optimized for use with Leica ST5010 and Leica ST5020, resulting in high quality H&E stains and consistent quality from the first stained slide to the last. High throughput. Time saving, as all stains and reagents exhaust at the same rate, allowing all components of the kit to be loaded on and off the instrument at the same time.
Catalog No. NC1231542 Supplier Leica Biosystems Supplier No. 3801698
All stains and reagents are optimized for use with Leica ST5010 and Leica ST5020, resulting in high quality H&E stains and consistent quality from the first stained slide to the last. High throughput. Time saving, as all stains and reagents exhaust at the same rate, allowing all components of the kit to be loaded on and off the instrument at the same time.