Learn More
Fine Science Tools S&T Vascular Clamps, Jaw: 5.5 x 1.5mm, 5 to 15 g, 11 mm
SDP
Catalog No. NC9314103
Encompass
Product: S&T Vascular Clamp - Straight/11mm/ Shape: Straight/ Jaw Dimensions: 5.5 x 1.5mm/ Clamping Pressure: Variable/ Gram Pressure: 5 to 15g/ Alloy: Stainless Steel/ Length: 11mm/ Serrated: No/ Unit: Each
Catalog No. NC9314103 Supplier Fine Science Tools Supplier No. 0039802
Product: S&T Vascular Clamp - Straight/11mm/ Shape: Straight/ Jaw Dimensions: 5.5 x 1.5mm/ Clamping Pressure: Variable/ Gram Pressure: 5 to 15g/ Alloy: Stainless Steel/ Length: 11mm/ Serrated: No/ Unit: Each