Learn More
Teknova Terrific Broth Complete with 1.6% Glycerol. 1000mL, Sterile. 2 pack.
SDP
Catalog No. NC2883725
Encompass
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.
Catalog No. NC2883725 Supplier Teknova Supplier No. T76612PK
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.