Learn More
ATCC ST; Testis; Pig (Sus scrofa)
Supplier:  ATCC CRL1746
Encompass
ST is a fibroblast-like cell that was isolated from the testis of a male pig. This cell line was deposited by SJ Wessman.
Catalog No. 50-238-1703