Learn More
Medchemexpress LLC HY-101279 5mg Medchemexpress, ST034307 CAS:133406-29-8 Purity:>98%
SDP
Catalog No. 501932087
Encompass_Preferred
Medchemexpress, HY-101279 5mg ST034307 CAS:133406-29-8 ST034307 is a potent and selective adenylyl cyclase 1 (AC1) inhibitor, with IC 50 of 2.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2087 Supplier Medchemexpress LLC Supplier No. HY1012795MG
Medchemexpress, HY-101279 5mg ST034307 CAS:133406-29-8 ST034307 is a potent and selective adenylyl cyclase 1 (AC1) inhibitor, with IC 50 of 2.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.