Learn More
Aviva Systems Biology Corp ST14 Antibody - middle region (ARP48057_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48057_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST14. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ST14. Alternate names: HAI, MT-SP1, MTSP-1, MTSP1, PRSS14, SNC19, TADG-15, TADG15, TMPRSS14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-342