Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ST1936 | 1210-81-7 | MFCD22201006 | 99.7% | 236.74 g/mol | C13H17ClN2 | 25 MG
SDP

Catalog No. 5000246016
Encompass_Preferred

ST1936 (CAS 1210-81-7) is a selective, nanomolar-affinity 5-HT6 receptor agonist used in pharmacological research. It is supplied as a solid with molecular formula C13H17ClN2, molecular weight 236.74 g/mol, and high reported purity suitable for in vitro studies.

  • High purity (99.7%) suitable for analytical and biological assays.
  • Selective for 5-HT6 receptors with nanomolar affinity.
  • Supplied as a 25 mg solid for small-scale laboratory use.
  • Molecular weight 236.74 g/mol and formula C13H17ClN2.
  • CAS number 1210-81-7 for unambiguous identification.

Catalog No. 50-002-46016 Supplier Medchemexpress LLC Supplier No. HY10311025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ST1936 (CAS 1210-81-7) is a selective, nanomolar-affinity 5-HT6 receptor agonist used in pharmacological research. It is supplied as a solid with molecular formula C13H17ClN2, molecular weight 236.74 g/mol, and high reported purity suitable for in vitro studies.

  • High purity (99.7%) suitable for analytical and biological assays.
  • Selective for 5-HT6 receptors with nanomolar affinity.
  • Supplied as a 25 mg solid for small-scale laboratory use.
  • Molecular weight 236.74 g/mol and formula C13H17ClN2.
  • CAS number 1210-81-7 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.