Learn More
Genscript Corporation ST2 8H6 mAb Mouse
Supplier:  Genscript Corporation V0610210
Encompass
Suppression of Tumorigenicity 2 (ST2) a member of the Interleukin-1 receptor family has two isoforms one is soluble form (sST2) and the other one is transmembrane form (ST2L) sST2 lacking the transmembrane and intracellular domains can prevent the interaction between IL-33 and ST2L It plays a role in inflammatory and immune processes and is considered as a promising marker for myocardial stress
Catalog No. 50-000-58014