Learn More
AdipoGen ST2 humanFc human rec
SDP
Catalog No. 502563029
Encompass_Preferred
Protein. Signal peptide and the sequence of isoform 2 of human ST2 aa 1-328 are fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS, pH 7.2. Binds to human IL-33. Purity >90% SDS-PAGE. The ST2 Interleukin-1 receptor-like 1 Interleukin-33 receptor gene was originally identified as a gene induced by serum or oncogene expression in fibroblasts. The gene produces a shorter soluble secreted form ST2 and a longer, transmembrane form ST2L by alternative splicing. Soluble ST2 has been shown to downregulate the expression of TLR1 and TLR4. ST2L negatively regulates TLR4 signaling and induces endotoxin tolerance, and enhances Th2 responses. IL-33 is the specific ligand for ST2L.
Catalog No. 50-256-3029 Supplier AdipoGen Supplier No. AG40A0059C050
Protein. Signal peptide and the sequence of isoform 2 of human ST2 aa 1-328 are fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS, pH 7.2. Binds to human IL-33. Purity >90% SDS-PAGE. The ST2 Interleukin-1 receptor-like 1 Interleukin-33 receptor gene was originally identified as a gene induced by serum or oncogene expression in fibroblasts. The gene produces a shorter soluble secreted form ST2 and a longer, transmembrane form ST2L by alternative splicing. Soluble ST2 has been shown to downregulate the expression of TLR1 and TLR4. ST2L negatively regulates TLR4 signaling and induces endotoxin tolerance, and enhances Th2 responses. IL-33 is the specific ligand for ST2L.