Learn More
Novus Biologicals™ ST2/IL-33R Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 9173 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-24706. For further blocking peptide related protocol, click here . |
| Gene Alias | DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed |
| Gene Symbol | IL1RL1 |
| Quantity | 0.1 mg |
| Research Category | Angiogenesis |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.