Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse ST2 (IL-33R) Uncoated ELISA Kit

Catalog No. 88933488
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-933-488 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-933-488 Supplier Invitrogen™ Supplier No. 88933488
Add to Cart
Add to Cart

ELISA

The Mouse ST-2 (IL-33R) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-33R. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-33R, also known as ST2, is a receptor for IL-33. It is a member of the IL-1 receptor family of innate receptors. Its extracellular domain consists of 3 Ig-like domains that directly interact with its co-receptor IL-1RAcP, and the ligand alarmin, IL-33. IL-33 ligation results in the association of IL-33R complex with its adaptor proteins MyD88, IRAK1, IRAK4 and TRAF6, and activation of the NF-kB, and MAPK pathways. This, in turn, instigates the release of chemokines and cytokines such as IL-5, IL-6, IL-8, and IL-13. Two most common isoforms of IL-33R that result from alternative splicing are ST2L - the membrane bound form, and ST2S - the soluble form. The membrane bound form can be released from the cell surface by proteolytic cleavage. The soluble forms have been suggested to act as decoys dampening the IL-33 signaling. IL-33R is primarily expressed by mast cells, basophils, and eosinophils. In mouse, IL-33R expression has been shown on ILC2 and Th2 cells. IL-33R signaling has been implicated in the number of immune conditions, including allergies, arthritis, atherosclerosis and sepsis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14719
Assay Range 80-10000 pg/mL
Assay Sensitivity 80 pg/mL
Content And Storage Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant protein for generating standard curve and calibrating samples
Coating Buffer: 10X PBS ELISA Coating Buffer
Assay Diluent: 5X Concentrated
Sample Diluent B: 12 mL of a 1X solution per plate
Detection Enzyme: Pre-titrated Avidin-HRP
Substrate Solution: Tetramethylbenzidine (TMB) Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards.
96-well Plates: Corning Costar flat-bottom plates (included with Cat. Nos. ending in -22 and -86)
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit
Product Type ELISA
Sample Type Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2; C87042; IL-17F;Ctla-8; Ctla8; IL-17; IL-17A; Il17
Gene ID (Entrez) 17082
Gene Symbol IL1RL1
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, Coating Buffer: 10X PBS ELISA Coating Buffer, Assay Diluent: 5X Concentrated, Sample Diluent B: 12 mL of a 1X solution per plate, Detection Enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22 and -86), Purified antibody, Detection Antibody: Pre-titrated, Biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, Coating Buffer: 10X PBS ELISA Coating Buffer, Assay Diluent: 5X Concentrated, Sample Diluent B: 12 mL of a 1X solution per plate, Detection Enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22 and -86)
Label or Dye Biotin
Protein Aliases Interleukin-33 receptor alpha chain, Lymphocyte antigen 84, Protein ST2, Protein T1
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 50 μL, Supernatant 50 μL
Storage Requirements 2-8°C
Synonym IL1RL1, Interleukin-1 Receptor-Like 1, Ly84
Target ST2
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
What are the critical parameters for successful ELISPOT assay?

A: It is critically important to use a high-affinity binding PVDF membrane plate, rather than regular ELISA plate or nitrocellulose membrane plate. Additionally, highest affinity antibodies for capture work best in this assay. Empirical optimization of conditions such as cell density, stimulus/mitogen, and kinetics is necessary.

I recently bought one of your Ready-Set-Go! ELISA Sets, can I use the antibody pairs to run an ELISPOT?

We do not recommend the use of our ELISA kits for the ELISPOT application. We cannot guarantee the successful use of our ELISA kits in an ELISPOT application.

Do your Ebioscience antibody pairs work in ELISPOT as well?

Most, but not all, of our ELISA antibody pairs work for ELISPOT. We continue to evaluate and optimize our anti-human and anti-mouse cytokine antibody pairs for ELISPOT applications. Please see our protocols page (www.thermofisher.com/us/en/home/references/protocols/cell-and-tissue-analysis/protocols) for a detailed protocol and a reference table with the list of antibody pairs that can be used in ELISPOT.

What are the components of the Ready-Set-Go! ELISPOT Sets?

Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
10X Sterile Azide-Free Coating Buffer
5X ELISA/ELISPOT Diluent
Detection enzyme: pre-titrated Avidin HRP
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and enzyme

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.