Learn More
Medchemexpress LLC HY-103097 5mg Medchemexpress, ST271 CAS:106392-48-7 Purity:>98%
SDP
Catalog No. 501968034
Encompass_Preferred
Medchemexpress, HY-103097 5mg ST271 CAS:106392-48-7 ST271 is a potent inhibitor of protein tyrosine kinase (PTK), inhibits phospholipase D activation stimulated by fMet-Leu-Phe and PAF, with IC50s of 6.7 and 9 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8034 Supplier Medchemexpress LLC Supplier No. HY1030975MG
Medchemexpress, HY-103097 5mg ST271 CAS:106392-48-7 ST271 is a potent inhibitor of protein tyrosine kinase (PTK), inhibits phospholipase D activation stimulated by fMet-Leu-Phe and PAF, with IC50s of 6.7 and 9 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.