Learn More
Aviva Systems Biology Corp ST3GAL2 Antibody - C-terminal region (ARP46800_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46800_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST3GAL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ST3GAL2. Alternate names: Gal-NAc6S, SIAT4B, ST3GALII, ST3GalA.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59404