Learn More
Aviva Systems Biology Corp ST3GAL4 Antibody - middle region (ARP46703_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46703_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST3GAL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ST3GAL4. Alternate names: CGS23, FLJ11867, NANTA3, SAT3, SIAT4, SIAT4C, ST3Gal IV, ST3GalIV, STZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-59377