Learn More
Aviva Systems Biology Corp ST3GAL5 Antibody - N-terminal region (ARP46385_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46385_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ST3GAL5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ST3GAL5. Alternate names: SIAT9, SIATGM3S, ST3GalV, SATIApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53475