Learn More
Aviva Systems Biology Corp St6gal2 Antibody - C-terminal region (ARP50078_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50078_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against St6gal2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C230064G14Rik, St6galII, mKIAA1877ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-482