Learn More
Aviva Systems Biology Corp St6galnac1 Antibody - C-terminal region (ARP47445_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47445_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against St6galnac1. It was validated on Western Blot. . Alternate names: Siat7aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-307